accessibility menu, dialog, popup

UserName

CAS RN 52462-29-0

CH3CH3H3CCH3CH3H3CClRuClClRuCl

CAS RN 52462-29-0

IUPAC Name: bis(1-methyl-4-(propan-2-yl)benzene); bis(dichlororuthenium)
Synonyms: (cymene)ruthenium dichloride dimer
Molecular Weight (g/mol): 612.38
Molecular Formula: C20H28Cl4Ru2
InChi Key: LAXRNWSASWOFOT-UHFFFAOYSA-J
SMILES: Cl[Ru]Cl.Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.CC(C)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)