CAS RN 52130-25-3
CAS RN 52130-25-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Hydroxy Dantrolene, TRC
CAS: 52130-25-3 Molecular Formula: C14 H10 N4 O6 Molecular Weight (g/mol): 330.25 IUPAC Name: 5-hydroxy-1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione SMILES: OC1N(\N=C\c2oc(cc2)c3ccc(cc3)[N+](=O)[O-])C(=O)NC1=O