accessibility menu, dialog, popup

UserName

NH2ONHNHO

CAS RN 521-31-3

IUPAC Name: 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione
Synonyms: luminol
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: HWYHZTIRURJOHG-UHFFFAOYSA-N
SMILES: NC1=C2C(=O)NNC(=O)C2=CC=C1
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (7)
  • (3)
Physical Form 
  • (4)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (7)
  • (3)
  • (4)
  • (2)
  • (4)