accessibility menu, dialog, popup

UserName

CAS RN 52093-27-3

IUPAC Name: praseodymium(3+) tritrifluoromethanesulfonate
Synonyms: praseodymium(3+) tritriflate
Molecular Weight (g/mol): 588.1
Molecular Formula: C3F9O9PrS3
InChi Key: ROUBZIWQWFQCHU-UHFFFAOYSA-K
SMILES: [Pr+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results