CAS RN 5204-74-0
IUPAC Name:
acetic acid; triethylamine
Synonyms:
triethylammonium acetate
Molecular Weight (g/mol):
161.25
Molecular Formula:
C8H19NO2
InChi Key:
AVBGNFCMKJOFIN-UHFFFAOYSA-N
SMILES:
CC(O)=O.CCN(CC)CC
1
–
2
of
2
results
1
Triethylammonium acetate buffer, BioUltra, volatile buffer, ∼1.0 M in H2O, Solstice
BioUltra, volatile buffer, ∼1.0 M in H2O
2
Acetic Acid – Triethylamine Solution 1:1, Honeywell Fluka™
CAS: 5204-74-0 Molecular Formula: C8H19NO2 Molecular Weight (g/mol): 161.245 MDL Number: MFCD00151725 InChI Key: AVBGNFCMKJOFIN-UHFFFAOYSA-N Synonym: triethylammonium acetate,teaa,acetic acid, compd. with n,n-diethylethanamine 1:1,acetic acid; n,n-diethylethanamine,ethanamine, n,n-diethyl-, acetate,triethylamine acetate,hoac et3n,acetic acid triethylamine,acetic acid; triethylamine,triethylammonium acetate solution PubChem CID: 165295 IUPAC Name: acetic acid;N,N-diethylethanamine SMILES: CCN(CC)CC.CC(=O)O