accessibility menu, dialog, popup

UserName

CH3(R)O(S)(S)(S)OOH(R)(S)HO(S)O

CAS RN 52-39-1

IUPAC Name: (1S,3aS,3bS,9aR,9bS,10S,11aR)-10-hydroxy-1-(2-hydroxyacetyl)-9a-methyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-11a-carbaldehyde
Synonyms: aldosterone D-aldosterone aldocortene 18-aldocorticosterone aldocortin aldocorten aldosteronum elektrocortin aldosterona [inn-spanish] electrocortin
Molecular Weight (g/mol): 360.45
Molecular Formula: C21H28O5
InChi Key: PQSUYGKTWSAVDQ-ZVIOFETBSA-N
SMILES: C[C@]12CCC(=O)C=C1CC[C@H]1[C@@H]3CC[C@H](C(=O)CO)[C@]3(C[C@H](O)[C@H]21)C=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)