accessibility menu, dialog, popup

UserName

CAS RN 519056-60-1

ONSNH2

CAS RN 519056-60-1

IUPAC Name: 4-(morpholin-4-yl)benzene-1-carbothioamide
Synonyms: 4-(morpholin-4-yl)benzenecarbothioamide
Molecular Weight (g/mol): 222.31
Molecular Formula: C11H14N2OS
InChi Key: KOPFTYFPHHZQCH-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results