accessibility menu, dialog, popup

UserName

CAS RN 51811-82-6

IUPAC Name: 7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Synonyms: 7-amino-N,N-dimethylphenothiazin-3-iminium chloride
Molecular Weight (g/mol): 291.8
Molecular Formula: C14H14ClN3S
InChi Key: NALREUIWICQLPS-UHFFFAOYSA-N
SMILES: [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (2)
  • (2)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)