accessibility menu, dialog, popup

UserName

NONCl

CAS RN 51802-77-8

IUPAC Name: 3-benzyl-5-(chloromethyl)-1,2,4-oxadiazole
Synonyms: 3-benzyl-5-(chloromethyl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 208.65
Molecular Formula: C10H9ClN2O
InChi Key: RUERNNLQCZMFKW-UHFFFAOYSA-N
SMILES: ClCC1=NC(CC2=CC=CC=C2)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results