accessibility menu, dialog, popup

UserName

CAS RN 518-67-2

IUPAC Name: 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide
Synonyms: 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide
Molecular Weight (g/mol): 380.29
Molecular Formula: C20H18BrN3
InChi Key: MQOKYEROIFEEBH-UHFFFAOYSA-N
SMILES: [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Molecular Weight (g/mol) 
  • (1)
  • (8)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results