accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 517-23-7

IUPAC Name: 3-acetyloxolan-2-one
Synonyms: 3-acetyloxolan-2-one
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: OMQHDIHZSDEIFH-UHFFFAOYNA-N
SMILES: CC(=O)C1CCOC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)