accessibility menu, dialog, popup

UserName

H3CCH3NH2Br

CAS RN 51605-97-1

IUPAC Name: 2-bromo-4-(propan-2-yl)aniline
Synonyms: 2-bromo-4-isopropylaniline
Molecular Weight (g/mol): 214.11
Molecular Formula: C9H12BrN
InChi Key: WEMDUNBELVTSRP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(N)C(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results