accessibility menu, dialog, popup

UserName

ONOClSClNOO

CAS RN 51584-21-5

IUPAC Name: 2,5-dichloro-3,4-dinitrothiophene
Synonyms: 2,5-dichloro-3,4-dinitrothiophene
Molecular Weight (g/mol): 243.01
Molecular Formula: C4Cl2N2O4S
InChi Key: UNRSLHGBQKYYSF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)SC(Cl)=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results