accessibility menu, dialog, popup

UserName

CAS RN 515832-52-7

CH3H3COBrN

CAS RN 515832-52-7

IUPAC Name: 5-bromo-2-(propan-2-yloxy)benzonitrile
Synonyms: 5-bromo-2-isopropoxybenzonitrile
Molecular Weight (g/mol): 240.1
Molecular Formula: C10H10BrNO
InChi Key: YRPPYKWHWKFYGJ-UHFFFAOYSA-N
SMILES: CC(C)OC1=C(C=C(Br)C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results