accessibility menu, dialog, popup

UserName

CAS RN 5144-89-8

IUPAC Name: 1,10-phenanthroline hydrate
Synonyms: 1,10-phenanthroline hydrate
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: PPQJCISYYXZCAE-UHFFFAOYSA-N
SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Quantity 
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (4)
  • (4)
  • (4)
Grade 
  • (2)
  • (4)
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (1)
  • (4)
  • (3)
  • (4)