CAS RN 5137-55-3
IUPAC Name:
methyltrioctylazanium chloride
Synonyms:
aliquat 336
Molecular Weight (g/mol):
404.16
Molecular Formula:
C25H54ClN
InChi Key:
XKBGEWXEAPTVCK-UHFFFAOYSA-M
SMILES:
[Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
1
–
2
of
2
results
1
Methyltrioctylammonium Chloride, TRC
CAS: 5137-55-3 Molecular Formula: C25 H54 N . Cl Molecular Weight (g/mol): 404.16 Synonym: Methyltrioctylammonium Chloride,A 336,A 336 (catalyst),Aliquat 128,Capriquat,Methyltri-n-octylammonium Chloride,Methyltricaprylammonium Chloride,N-Methyl-N,N,N-trioctylammonium Chloride,TOMAC,TOMAC 75,Tri-n-octylmethylammonium Chloride,Tricaprylmethylammonium Chloride,Trioctylmonomethylammonium Chloride; IUPAC Name: methyl(trioctyl)azanium;chloride SMILES: [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
2
Trioctylmethylammonium Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.