accessibility menu, dialog, popup

UserName

CAS RN 51142-49-5

FFFFFNFFFFFFFFFFFFFFFFFFFFFFFFFFFNFFFFFFFFFFFFFFFF

CAS RN 51142-49-5

IUPAC Name: bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)(trifluoromethyl)amine; tris(1,1,2,2,3,3,4,4,4-nonafluorobutyl)amine
Synonyms: bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)(trifluoromethyl)amine; perfluorotributylamine
Molecular Weight (g/mol): 1192.17
Molecular Formula: C21F48N2
InChi Key: QDOIZVITZUBGOQ-UHFFFAOYSA-N
SMILES: FC(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)