accessibility menu, dialog, popup

UserName

CAS RN 5111-65-9

IUPAC Name: 2-bromo-6-methoxynaphthalene
Synonyms: 2-bromo-6-methoxynaphthalene
Molecular Weight (g/mol): 237.1
Molecular Formula: C11H9BrO
InChi Key: AYFJBMBVXWNYLT-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(Br)C=CC2=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)