accessibility menu, dialog, popup

UserName

CAS RN 51047-59-7

CH3NNNHN

CAS RN 51047-59-7

IUPAC Name: 2-methyl-6-(piperazin-1-yl)pyrazine
Synonyms: 2-methyl-6-(piperazin-1-yl)pyrazine
Molecular Weight (g/mol): 178.24
Molecular Formula: C9H14N4
InChi Key: ZQAVHWZTBHGKFK-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=N1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results