CAS RN 51-61-6
CAS RN 51-61-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dopamine, TRC
CAS: 51-61-6 Molecular Formula: C8 H11 N O2 Molecular Weight (g/mol): 153.18 Synonym: 1,2-Benzenediol, 4-(2-aminoethyl)-,Pyrocatechol, 4-(2-aminoethyl)- (8CI),4-(2-Aminoethyl)-1,2-benzenediol,2-(3,4-Dihydroxyphenyl)-1-ethanamine,2-(3,4-Dihydroxyphenyl)ethylamine,3,4-Dihydroxyphenethylamine,3,4-Dihydroxyphenylethylamine,3-Hydroxytyramine,4-(2-Aminoethyl)catechol,4-(2-Aminoethyl)pyrocatechol,DA,Dopamin,Dopamine,Dophamine,Duobaan,Hydroxytyramin,NSC 173182,Oxytyramine,α-(3,4-Dihydroxyphenyl)-β-aminoethane IUPAC Name: 4-(2-aminoethyl)benzene-1,2-diol SMILES: NCCc1ccc(O)c(O)c1