accessibility menu, dialog, popup

UserName

CAS RN 51-17-2

HNN

CAS RN 51-17-2

IUPAC Name: 1H-1,3-benzodiazole
Synonyms: benzimidazole azindole benziminazole BZI benzimidazole (substance) Hbim benzoglyoxaline hbzim
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: HYZJCKYKOHLVJF-UHFFFAOYSA-N
SMILES: N1C=NC2=CC=CC=C12
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)