accessibility menu, dialog, popup

UserName

CAS RN 50921-39-6

OOCl

CAS RN 50921-39-6

IUPAC Name: 1-(4-chlorophenyl)cyclobutane-1-carboxylate
Synonyms: 1-(4-chlorophenyl)cyclobutane-1-carboxylate
Molecular Weight (g/mol): 209.65
Molecular Formula: C11H10ClO2
InChi Key: XYSRHOKREWGGFE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1(CCC1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results