accessibility menu, dialog, popup

UserName

CAS RN 50910-54-8

NH2OHHCl

CAS RN 50910-54-8

IUPAC Name: 4-aminocyclohexan-1-ol hydrochloride
Synonyms: 4-aminocyclohexan-1-ol hydrochloride
Molecular Weight (g/mol): 151.63
Molecular Formula: C6H14ClNO
InChi Key: RKTQEVMZBCBOSB-UHFFFAOYSA-N
SMILES: Cl.NC1CCC(O)CC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results