accessibility menu, dialog, popup

UserName

CH3OONSH2N

CAS RN 50850-93-6

IUPAC Name: ethyl 2-amino-1,3-benzothiazole-6-carboxylate
Synonyms: ethyl 2-amino-1,3-benzothiazole-6-carboxylate
Molecular Weight (g/mol): 222.26
Molecular Formula: C10H10N2O2S
InChi Key: VYJSGJXWKSDUSG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C2N=C(N)SC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results