accessibility menu, dialog, popup

UserName

H3CCH3(S)(S)CH3(R)(R)H3C(S)OHCH3H3C(R)(R)CH3(S)OHO

CAS RN 508-02-1

IUPAC Name: (4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
Synonyms: oleanolic acid hederins astrantiagenin C virgaureagenin B oleanol oleanic acid giganteumgenin C triterpenes, oleanane oleanane triterpenes caryophyllin
Molecular Weight (g/mol): 456.71
Molecular Formula: C30H48O3
InChi Key: MIJYXULNPSFWEK-GTOFXWBISA-N
SMILES: CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (7)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)