accessibility menu, dialog, popup

UserName

CAS RN 50715-50-9

IUPAC Name: 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3-{[(tert-butoxy)carbonyl]amino}butanedioate
Synonyms: 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3-[(tert-butoxycarbonyl)amino]butanedioate
Molecular Weight (g/mol): 386.4
Molecular Formula: C17H26N2O8
InChi Key: PIITZDSTZQZNQH-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC(NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results