accessibility menu, dialog, popup

UserName

CAS RN 50673-97-7

H3CCH3CH3(R)O(S)(S)OH(S)H3C(R)H3C

CAS RN 50673-97-7

IUPAC Name: (1R,5aS,7S,9aS,11aR)-7-hydroxy-9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,4H,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-3-one
Synonyms: (1R,5aS,7S,9aS,11aR)-7-hydroxy-9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,4H,5H,5aH,6H,7H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol): 400.65
Molecular Formula: C27H44O2
InChi Key: LINVVMHRTUSXHL-NDNSGUFDSA-N
SMILES: CC(C)CCCC(C)[C@H]1CC(=O)C2=C3CC[C@H]4C[C@@H](O)CC[C@]4(C)C3CC[C@]12C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results