accessibility menu, dialog, popup

UserName

CAS RN 50670-49-0

IUPAC Name: 4-bromo-4'-methyl-1,1'-biphenyl
Synonyms: 4-bromo-4'-methyl-1,1'-biphenyl
Molecular Weight (g/mol): 247.14
Molecular Formula: C13H11Br
InChi Key: MYWLXURJCXISCT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results