accessibility menu, dialog, popup

UserName

CAS RN 50607-30-2

IUPAC Name: piperidine-2,4-dione
Synonyms: piperidine-2,4-dione
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: RDNZDMDLRIQQAX-UHFFFAOYSA-N
SMILES: O=C1CCNC(=O)C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results