accessibility menu, dialog, popup

UserName

NH3NH3OHOHO

CAS RN 506-87-6

IUPAC Name: carbonic acid diamine
Synonyms: ammonium carbonate
Molecular Weight (g/mol): 96.09
Molecular Formula: CH8N2O3
InChi Key: PRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILES: N.N.OC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (19)
  • (19)
Quantity 
  • (9)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (10)
  • (9)
Grade 
  • (2)
  • (8)
  • (4)
  • (2)
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
  • (7)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (5)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (7)
  • (2)
  • (19)
  • (19)
  • (9)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (4)
  • (10)
  • (9)
  • (2)
  • (8)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (5)
  • (7)
  • (2)
  • (2)