CAS RN 506-78-5
CAS RN 506-78-5
IUPAC Name:
carbononitridic iodide
Synonyms:
cyanogen iodide
Molecular Weight (g/mol):
152.92
Molecular Formula:
CIN
InChi Key:
WPBXOELOQKLBDF-UHFFFAOYSA-N
SMILES:
IC#N
1
–
1
of
1
results
1
Promotions Available
Cyanogen iodide, 98%, Thermo Scientific Chemicals
Cyanogen iodide, 98%, Thermo Scientific Chemicals
CAS: 506-78-5 Molecular Formula: CIN InChI Key: WPBXOELOQKLBDF-UHFFFAOYSA-N Synonym: cyanogen iodide,iodine cyanide,jodcyan,iodocyanide,iodine monocyanide,cyanogen monoiodide,iodine cyanide i cn,unii-81esv254uf,cyanic iodide,cyano iodide PubChem CID: 10478 IUPAC Name: carbononitridic iodide SMILES: C(#N)I