accessibility menu, dialog, popup

UserName

CAS RN 506-61-6

KAgCNCN

CAS RN 506-61-6

IUPAC Name: potassium silver(1+) bis(iminomethanide)
Synonyms: potassium argentocyanide
Molecular Weight (g/mol): 199
Molecular Formula: C2AgKN2
InChi Key: HKSGQTYSSZOJOA-UHFFFAOYSA-N
SMILES: [K+].[Ag+].[C-]#N.[C-]#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)