CAS RN 506-21-8
IUPAC Name:
(9Z,12Z)-octadeca-9,12-dienoic acid
Synonyms:
linoleic pure linoleic acid acide linoleique oils, grape oils, grape seed emersol 315 emersol 310 polylin no. 515 acide cis-linoleique grape seed oil
Molecular Weight (g/mol):
280.45
Molecular Formula:
C18H32O2
InChi Key:
OYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILES:
CCCCC\C=C/C\C=C/CCCCCCCC(O)=O
1
–
1
of
1
results
1
(9E,12E)-Octadeca-9,12-dienoic Acid, TRC
CAS: 506-21-8 Molecular Formula: C18 H32 O2 Molecular Weight (g/mol): 280.45 Synonym: Linolelaidic Acid,9,12-Octadecadienoic acid, (E,E)-,Linolelaidic acid (8CI),(9E,12E)-9,12-Octadecadienoic acid,9-trans-12-trans-Linoleic acid,9-trans-12-trans-Octadecadienoic acid,C18:2n-6(t9,t12),Linelaidic acid,Linoelaidic acid,trans,trans-9,12-Octadecadienoic acid,trans,trans-Linoleic acid,trans-9-trans-12-Linoleic acid,trans-δ9,12-Octadecadienoic acid,9,12-Octadecadienoic acid, (9E,12E)- IUPAC Name: (9E,12E)-octadeca-9,12-dienoic acid SMILES: CCCCC\C=C\C\C=C\CCCCCCCC(=O)O