CAS RN 50594-82-6
IUPAC Name:
1,2,3-trichloro-5-(trifluoromethyl)benzene
Synonyms:
1,2,3-trichloro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol):
249.44
Molecular Formula:
C7H2Cl3F3
InChi Key:
FBKFIAIRSQOXJR-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(Cl)=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
3,4,5-Trichlorobenzotrifluoride, 97%, Thermo Scientific™
CAS: 50594-82-6 Molecular Formula: C7H2Cl3F3 Molecular Weight (g/mol): 249.438 MDL Number: MFCD00035985 InChI Key: FBKFIAIRSQOXJR-UHFFFAOYSA-N Synonym: 1,2,3-trichloro-5-trifluoromethyl benzene,3,4,5-trichlorobenzotrifluoride,3,4,5-trichlorotrifluoromethylbenzene,3,4,5-trichloro-alpha,alpha,alpha-trifluorotoluene,3,4,5-trichloro-a,a,a-trifluorotoluene,benzene, 1,2,3-trichloro-5-trifluoromethyl,pubchem19737,ksc914i9j,3,4,5-trichloro benzotrifluoride PubChem CID: 2733399 IUPAC Name: 1,2,3-trichloro-5-(trifluoromethyl)benzene SMILES: C1=C(C=C(C(=C1Cl)Cl)Cl)C(F)(F)F