accessibility menu, dialog, popup

UserName

CAS RN 5059-36-9

CH3H3CNNHN

CAS RN 5059-36-9

IUPAC Name: 1-[2-(2,5-dimethyl-1H-pyrrol-1-yl)ethyl]piperazine
Synonyms: 1-[2-(2,5-dimethylpyrrol-1-yl)ethyl]piperazine
Molecular Weight (g/mol): 207.32
Molecular Formula: C12H21N3
InChi Key: AUSXFRFXXDHSTC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1CCN1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results