CAS RN 5057-98-7
CAS RN 5057-98-7
IUPAC Name:
cyclopentane-1,2-diol
Synonyms:
cyclopentane-1,2-diol
Molecular Weight (g/mol):
102.13
Molecular Formula:
C5H10O2
InChi Key:
VCVOSERVUCJNPR-UHFFFAOYNA-N
SMILES:
OC1CCCC1O
1
–
1
of
1
results
1
Promotions Available
cis-1,2-Cyclopentanediol, 98%
cis-1,2-Cyclopentanediol, 98%
CAS: 5057-98-7 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 MDL Number: MFCD00134192 InChI Key: VCVOSERVUCJNPR-SYDPRGILSA-N Synonym: cis-1,2-cyclopentanediol,cis-cyclopentane-1,2-diol,cis-1,2-dihydroxycyclopentane,1r,2s-cyclopentane-1,2-diol,1s,2r cyclopentane-1,2-diol,cis-1,2-cyclopentandiol,1,2-cis-cyclopentanediol,cis-1,2-cyclopen-tanediol,cis-cyclopenten-1,2-diol,1alpha,2alpha-cyclopentanediol PubChem CID: 2733293 IUPAC Name: (1R,2S)-cyclopentane-1,2-diol SMILES: C1CC(C(C1)O)O