accessibility menu, dialog, popup

UserName

NH2N

CAS RN 50541-93-0

IUPAC Name: 1-benzylpiperidin-4-amine
Synonyms: 1-benzylpiperidin-4-amine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: YUBDLZGUSSWQSS-UHFFFAOYSA-N
SMILES: NC1CCN(CC2=CC=CC=C2)CC1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)