accessibility menu, dialog, popup

UserName

CAS RN 505084-59-3

CAS RN 505084-59-3

IUPAC Name: 2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
Synonyms: 2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: RMSKYTKOFDQVEL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CN=C1Cl)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)