accessibility menu, dialog, popup

UserName

CAS RN 5044-22-4

CH3H3CNNOO

CAS RN 5044-22-4

IUPAC Name: 2,5-dimethyl-1-(4-nitrophenyl)-1H-pyrrole
Synonyms: 2,5-dimethyl-1-(4-nitrophenyl)pyrrole
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: JXJRCFOGAIXRCU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results