CAS RN 504-88-1
CAS RN 504-88-1
IUPAC Name:
3-nitropropanoic acid
Synonyms:
β-nitropropionic acid bovinocidin
Molecular Weight (g/mol):
119.08
Molecular Formula:
C3H5NO4
InChi Key:
WBLZUCOIBUDNBV-UHFFFAOYSA-N
SMILES:
OC(=O)CC[N+]([O-])=O
1
–
1
of
1
results
1
3-Nitropropanoic acid, MedChemExpress
MedChemExpress 3-Nitropropanoic acid (β-Nitropropionic acid) is an irreversible inhibitor of succinate dehydrogenase. 3-Nitropropanoic acid exhibits potent antimycobacterial activity with a MIC value of 3.3 μM.