missing translation for 'onlineSavingsMsg'
Learn More

3-Nitropropanoic acid, MedChemExpress

Product Code. 16462506
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16462506

Brand: Medchem Express HYW012875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Nitropropanoic acid (β-Nitropropionic acid) is an irreversible inhibitor of succinate dehydrogenase. 3-Nitropropanoic acid exhibits potent antimycobacterial activity with a MIC value of 3.3 μM.

3-Nitropropanoic acid (β-Nitropropionic acid) is an irreversible inhibitor of succinate dehydrogenase. 3-Nitropropanoic acid exhibits potent antimycobacterial activity with a MIC value of 3.3 μM.

Specifications

Chemical Name or Material 3-Nitropropanoic acid
CAS 504-88-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C3H5NO4
Quantity 500 mg
Synonym β-Nitropropionic acid Bovinocidin
Solubility Information DMSO : 125 mg/mL (1049.71 mM; Need ultrasonic)
SMILES O=C(O)CC[N+]([O-])=O
Molecular Weight (g/mol) 119.08
Formula Weight 119.08
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.