accessibility menu, dialog, popup

UserName

H2NNOHCl(Z)

CAS RN 5033-28-3

IUPAC Name: (Z)-4-chloro-N'-hydroxybenzene-1-carboximidamide
Synonyms: (Z)-4-chloro-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: QBGONPQFBDUVPG-UHFFFAOYSA-N
SMILES: N\C(=N/O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)