accessibility menu, dialog, popup

UserName

CAS RN 502-44-3

OO

CAS RN 502-44-3

IUPAC Name: oxepan-2-one
Synonyms: caprolactone E-caprolactone 2-oxohexamethylene oxide 1,6-hexanolide 6-hydroxyhexanoic acid lactone 6-hexanolactone epsilon-lactone hexanoic acid hexamethylene oxide, 2-oxo- epsilon-kaprolakton hexan-6-olide
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: PAPBSGBWRJIAAV-UHFFFAOYSA-N
SMILES: O=C1CCCCCO1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (7)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (4)
  • (4)
  • (2)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (3)
  • (4)
  • (4)