accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHOOHBr

CAS RN 501015-16-3

IUPAC Name: 3-(3-bromophenyl)-3-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-(3-bromophenyl)-3-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 344.21
Molecular Formula: C14H18BrNO4
InChi Key: UTYRIGRZLZNTLR-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)C1=CC(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results