accessibility menu, dialog, popup

UserName

CAS RN 501-94-0

OHOH

CAS RN 501-94-0

IUPAC Name: 4-(2-hydroxyethyl)phenol
Synonyms: tyrosol n-tyrosol p-tyrosol p-thyrosol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: YCCILVSKPBXVIP-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)