accessibility menu, dialog, popup

UserName

CAS RN 501-65-5

CAS RN 501-65-5

IUPAC Name: (2-phenylethynyl)benzene
Synonyms: tolan tolane biphenylacetylene
Molecular Weight (g/mol): 178.23
Molecular Formula: C14H10
InChi Key: JRXXLCKWQFKACW-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C#CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)