accessibility menu, dialog, popup

UserName

CAS RN 5004-45-5

OHNN

CAS RN 5004-45-5

IUPAC Name: 4-phenyl-1,2-dihydrophthalazin-1-one
Synonyms: 1(2H)-phthalazinone, 4-phenyl-
Molecular Weight (g/mol): 222.25
Molecular Formula: C14H10N2O
InChi Key: XCJLBNVENUPHEA-UHFFFAOYSA-N
SMILES: O=C1NN=C(C2=CC=CC=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results