accessibility menu, dialog, popup

UserName

CAS RN 5001-18-3

OHOH

CAS RN 5001-18-3

IUPAC Name: adamantane-1,3-diol
Synonyms: adamantane-1,3-diol
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: MOLCWHCSXCKHAP-UHFFFAOYSA-N
SMILES: OC12CC3CC(C1)CC(O)(C3)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results