CAS RN 5000-46-4
CAS RN 5000-46-4
IUPAC Name:
1-methanesulfonylpropan-2-one
Synonyms:
1-methanesulfonylpropan-2-one
Molecular Weight (g/mol):
136.17
Molecular Formula:
C4H8O3S
InChi Key:
NWEYGXQKFVGUFR-UHFFFAOYSA-N
SMILES:
CC(=O)CS(C)(=O)=O
1
–
1
of
1
results
1
Methylsulfonylacetone, 98+%
CAS: 5000-46-4 Molecular Formula: C4H8O3S Molecular Weight (g/mol): 136.165 MDL Number: MFCD00014745 InChI Key: NWEYGXQKFVGUFR-UHFFFAOYSA-N Synonym: methylsulfonylacetone,1-methylsulfonyl propan-2-one,methanesulfonylacetone,1-methylsulfonyl acetone,1-methanesulfonylpropan-2-one,1-methylsulphonyl acetone,2-propanone, 1-methylsulfonyl,methylsulfonylaceton,methylsulphonylacetone,methanesulphonylacetone PubChem CID: 78695 IUPAC Name: 1-methylsulfonylpropan-2-one SMILES: CC(=O)CS(=O)(=O)C