CAS RN 50-70-4
Thermo Scientific Chemicals D-Sorbitol, 97%
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00004708 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
Thermo Scientific Chemicals D-Sorbitol, 98%
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.172 MDL Number: MFCD00004708 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
Sorbitol, Fisher BioReagents
D-Sorbitol, Molecular Biology Reagent, ≥98%, MP Biomedicals™
CAS: 50-70-4 Molecular Formula: C6H14O6 Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924
D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98%, MP Biomedicals™
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,glucarine,diakarmon,multitol,sorbilande PubChem CID: 5780 ChEBI: CHEBI:17924
Melford D-Sorbitol
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Sorbitol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
D-Sorbitol, TRC
CAS: 50-70-4 Molecular Formula: C6 H14 O6 Molecular Weight (g/mol): 182.17 Synonym: Sorbitol,d-Glucitol,d-sorbitol IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO
D-Sorbit 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
D-Sorbitol, MedChemExpress
MedChemExpress D-Sorbitol (Sorbitol) is a six-carbon sugar alcohol and can used as a sugar substitute. D-Sorbitol can be used as a stabilizing excipient and/or isotonicity agent, sweetener, humectant, thickener and dietary supplement.
D-Sorbitol, 91.0 to 100.5% (Dry Basis), Ultrapure
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.172 MDL Number: MFCD00004708 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2 R,3 R,4 R,5 S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
D-Sorbitol, >98%, MP Biomedicals™
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.172 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
Sorbitol, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.172 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
- 1
- 2